Cyclopropyl-3-methoxyphenyl ketone ≥95%

Supplier: AOBChem USA

Warning

233199-5G 233199-25G 233199-1G 233199-500MG
77632-106EA 1161 USD
77632-106 77658-094 77641-322 77649-358
Cyclopropyl-3-methoxyphenyl ketone ≥95%
Cyclopropyl-3-methoxyphenyl ketone

Precaución: For Research Use Only

Formula: C₁₁H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03841209
CAS Number: 104271-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR