Cyclopropyl(3,4-dichlorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

233195-5G 233195-25G 233195-1G 233195-500MG
77632-104EA 765 USD
77632-104 77654-274 77641-320 77645-306
Cyclopropyl(3,4-dichlorophenyl)methanone ≥95%
Cyclopropyl(3,4-dichlorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD03841239
CAS Number: 136906-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR