1-(3,4-Dichlorophenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

233182-5G 233182-25G 233182-1G 233182-500MG
77632-102EA 827 USD
77632-102 77654-272 77641-318 77645-304
1-(3,4-Dichlorophenyl)-2-methylpropan-1-one ≥95%
1-(3,4-Dichlorophenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: C₁₀H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD09891220
CAS Number: 875916-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR