5-Bromo-2-ethoxy-1-fluoro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

23313-5G 23313-25G 23313-1G 23313-500MG
77632-100EA 933 USD
77632-100 77658-092 77641-316 77649-356
5-Bromo-2-ethoxy-1-fluoro-3-(trifluoromethyl)benzene ≥97%
5-Bromo-2-ethoxy-1-fluoro-3-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: C₉H₇BrF₄O
MW: 287.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056755
CAS Number: 1370025-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR