5-((Diethylamino)methyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

23020-5G 23020-1G 23020-25G 23020-500MG
77632-094EA 1117 USD
77632-094 77637-320 77654-264 77649-348
5-((Diethylamino)methyl)-2-fluorophenylboronic acid ≥97%
5-((Diethylamino)methyl)-2-fluorophenylboronic acid

Precaución: For Research Use Only

Formula: 225.068 g/mol
MW: C₁₁H₁₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD14687159
CAS Number: 1334226-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR