1-[6-(1-Methylethoxy)-2-pyridinyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

229803-5G 229803-1G 229803-25G 229803-500MG
77632-090EA 1130 USD
77632-090 77637-314 77654-260 77645-294
1-[6-(1-Methylethoxy)-2-pyridinyl]ethanone ≥95%
1-[6-(1-Methylethoxy)-2-pyridinyl]ethanone

Precaución: For Research Use Only

Formula: 179.22 g/mol
MW: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD23114016
CAS Number: 1394957-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR