2-Ethoxy-6-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

229675-5G 229675-25G 229675-1G 229675-500MG
77628-146EA 1526.46 USD
77628-146 77635-538 77637-310 77649-344
2-Ethoxy-6-fluoropyridine ≥97%
2-Ethoxy-6-fluoropyridine

Precaución: For Research Use Only

Formula: 141.14 g/mol
MW: C₇H₈FNO
Storage Temperature: Ambient
CAS Number: 858675-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR