4-(2-Methoxyphenyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

22899-5G 22899-1G 22899-500MG
77628-142EA 1484.23 USD
77628-142 77645-288 77649-340
4-(2-Methoxyphenyl)phenylboronic acid ≥95%
4-(2-Methoxyphenyl)phenylboronic acid

Precaución: For Research Use Only

Formula: C₁₃H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD08701729
CAS Number: 1107041-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR