Methyl 2,6-difluoro-4-ethoxybenxoate ≥97%

Supplier: AOBChem USA

Warning

22864-5G 22864-1G 22864-25G 22864-500MG
77634-420EA 507 USD
77634-420 77637-306 77654-254 77645-286
Methyl 2,6-difluoro-4-ethoxybenxoate ≥97%
Methyl 2,6-difluoro-4-ethoxybenxoate

Precaución: For Research Use Only

Formula: 216.181 g/mol
MW: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD18204755
CAS Number: 1351668-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR