4-Hydroxy-2,5-bis(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

228339-1G 228339-5G 228339-500MG
77634-418EA 1032 USD
77634-418 77654-252 77649-336
4-Hydroxy-2,5-bis(trifluoromethyl)benzaldehyde ≥95%
4-Hydroxy-2,5-bis(trifluoromethyl)benzaldehyde

Precaución: For Research Use Only

Formula: C₉H₄F₆O₂
Storage Temperature: Ambient
MDL Number: MFCD28730898
CAS Number: 1805588-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR