3-Bromo-6-hydroxy-2,4-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

228039-5G 228039-25G 228039-1G 228039-500MG
77632-084EA 870 USD
77632-084 77654-250 77641-300 77645-282
3-Bromo-6-hydroxy-2,4-dimethylbenzaldehyde ≥97%
3-Bromo-6-hydroxy-2,4-dimethylbenzaldehyde

Precaución: For Research Use Only

Formula: 229.07 g/mol
MW: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11500997
CAS Number: 1172886-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR