6-Bromo-3-chloro-2-hydroxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

228002-5G 228002-500MG 228002-25G 228002-1G
77632-082EA 1388 USD
77632-082 77652-508 77658-082 77641-298
6-Bromo-3-chloro-2-hydroxybenzaldehyde ≥95%
6-Bromo-3-chloro-2-hydroxybenzaldehyde

Precaución: For Research Use Only

Formula: C₇H₄BrClO₂
Storage Temperature: Ambient
CAS Number: 1427327-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR