2-Bromo-6-hydroxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

227995-5G 227995-1G 227995-25G 227995-500MG
77632-080EA 1044 USD
77632-080 77637-304 77658-080 77645-280
2-Bromo-6-hydroxy-3-methylbenzaldehyde ≥97%
2-Bromo-6-hydroxy-3-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD21603899
CAS Number: 1416373-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR