3-Bromo-2-fluoro-4-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

22794-5G 22794-1G 22794-25G 22794-500MG
77634-416EA 597 USD
77634-416 77637-302 77658-078 77645-278
3-Bromo-2-fluoro-4-methylbenzaldehyde ≥95%
3-Bromo-2-fluoro-4-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD22201081
CAS Number: 1373223-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR