2-Bromo-2'-fluoro-biphenyl ≥95%

Supplier: AOBChem USA

Warning

22730-5G 22730-1G 22730-25G 22730-500MG
77632-076EA 1216 USD
77632-076 77637-298 77654-246 77645-276
2-Bromo-2'-fluoro-biphenyl ≥95%
2-Bromo-2'-fluoro-biphenyl

Precaución: For Research Use Only

Formula: C₁₂H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD16659072
CAS Number: 1554-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR