3,5-Dichloro-4-methylaniline ≥97%

Supplier: AOBChem USA

Warning

22714-5G 22714-25G
77634-414EA 59 USD
77634-414 77658-076
3,5-Dichloro-4-methylaniline ≥97%
3,5-Dichloro-4-methylaniline

Precaución: For Research Use Only

Formula: C7H7Cl2N
Storage Temperature: Ambient
MDL Number: MFCD00266300
CAS Number: 54730-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR