4',5'-Dimethyl-2'-methoxyacetophenone ≥95%

Supplier: AOBChem USA

Warning

225893-5G 225893-25G 225893-1G
77634-408EA 214 USD
77634-408 77654-242 77641-296
4',5'-Dimethyl-2'-methoxyacetophenone ≥95%
4',5'-Dimethyl-2'-methoxyacetophenone

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789142
CAS Number: 91969-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR