2-(3,4-Dimethylphenoxy)ethanol ≥97%

Supplier: AOBChem USA

Warning

225892-5G 225892-500MG 225892-25G 225892-1G
77634-406EA 440 USD
77634-406 77652-504 77654-240 77641-294
2-(3,4-Dimethylphenoxy)ethanol ≥97%
2-(3,4-Dimethylphenoxy)ethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11613991
CAS Number: 34221-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR