2-Methoxy-4,5-dimethylphenol ≥95%

Supplier: AOBChem USA

Warning

225881-5G 225881-1G 225881-25G 225881-500MG
77632-074EA 1206 USD
77632-074 77637-292 77658-072 77649-334
2-Methoxy-4,5-dimethylphenol ≥95%
2-Methoxy-4,5-dimethylphenol

Precaución: For Research Use Only

Formula: 152.19 g/mol
MW: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD14538105
CAS Number: 7771-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR