2-Bromo-4,6-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

225695-5G 225695-25G 225695-1G 225695-500MG
77632-072EA 1226 USD
77632-072 77658-070 77641-292 77649-332
2-Bromo-4,6-dimethoxybenzoic acid ≥97%
2-Bromo-4,6-dimethoxybenzoic acid

Precaución: For Research Use Only

Formula: C₉H₉BrO₄
Storage Temperature: Ambient
MDL Number: MFCD18906166
CAS Number: 62827-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR