2-(4-(Benzyloxy)-3,5-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

22559-5G 22559-25G
77634-404EA 59 USD
77634-404 77658-068
2-(4-(Benzyloxy)-3,5-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-(Benzyloxy)-3,5-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For Research Use Only

Formula: C₁₉H₂₁BF₂O₃
MW: 346.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383455
CAS Number: 1255041-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR