1-Benzyl-7-bromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22553-5G 22553-1G 22553-25G 22553-500MG
77634-402EA 388 USD
77634-402 77637-290 77654-238 77645-274
1-Benzyl-7-bromo-1H-indazole ≥97%
1-Benzyl-7-bromo-1H-indazole

Precaución: For Research Use Only

Formula: C₁₄H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD20526381
CAS Number: 1351668-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR