5-Ethoxypyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

22546-5G 22546-1G 22546-500MG 22546-25G
77632-070EA 618 USD
77632-070 77637-288 77652-502 77658-066
5-Ethoxypyridine-3-carboxylic acid ≥97%
5-Ethoxypyridine-3-carboxylic acid

Precaución: For Research Use Only

Formula: 167.162 g/mol
MW: C₈H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD18898180
CAS Number: 98491-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR