4-Chloro-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22531-5G 22531-1G 22531-25G 22531-500MG
77632-068EA 950 USD
77632-068 77637-286 77658-062 77649-330
4-Chloro-2-fluoro-3-methylbenzaldehyde ≥97%
4-Chloro-2-fluoro-3-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD20526379
CAS Number: 1351668-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR