6-Fluoro-5-iodo-3-pyridinecarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

223892-1G 223892-500MG
77634-396EA 3380 USD
77634-396 77649-326
6-Fluoro-5-iodo-3-pyridinecarboxylic acid ≥95%
6-Fluoro-5-iodo-3-pyridinecarboxylic acid

Precaución: For Research Use Only

Formula: 267 g/mol
MW: C₆H₃FINO₂
Storage Temperature: Ambient
CAS Number: 1674401-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR