Benzoic acid, 2,5-dichloro-4-(methylsulfonyl)- 95

Supplier: AOBChem USA

Warning

223381-5G 223381-25G 223381-1G 223381-500MG
77632-060EA 688 USD
77632-060 77658-046 77641-284 77645-264
Benzoic acid, 2,5-dichloro-4-(methylsulfonyl)- 95
Benzoic acid, 2,5-dichloro-4-(methylsulfonyl)-

Precaución: For Research Use Only

Formula: C₈H₆Cl₂O₄S
MW: 269.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34869398
CAS Number: 118939-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR