4-Bromo-2-(methylsulfonyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

223269-5G 223269-25G 223269-1G 223269-500MG
77632-058EA 967 USD
77632-058 77654-234 77641-282 77649-322
4-Bromo-2-(methylsulfonyl)pyridine ≥95%
4-Bromo-2-(methylsulfonyl)pyridine

Precaución: For Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702650
CAS Number: 1209459-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR