1-Iodo-3-(methylsulfonyl)benzene ≥95%

Supplier: AOBChem USA

Warning

223127-5G 223127-1G 223127-500MG
77628-136EA 1484.23 USD
77628-136 77637-276 77645-260
1-Iodo-3-(methylsulfonyl)benzene ≥95%
1-Iodo-3-(methylsulfonyl)benzene

Precaución: For Research Use Only

Formula: C₇H₇IO₂S
Storage Temperature: Ambient
MDL Number: MFCD00955956
CAS Number: 139769-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR