1,4-Dimethyl-2-(methylsulfonyl)benzene ≥95%

Supplier: AOBChem USA

Warning

223123-5G 223123-1G 223123-25G 223123-500MG
77634-394EA 530 USD
77634-394 77637-274 77658-044 77645-258
1,4-Dimethyl-2-(methylsulfonyl)benzene ≥95%
1,4-Dimethyl-2-(methylsulfonyl)benzene

Precaución: For Research Use Only

Formula: C₉H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD03789176
CAS Number: 6462-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR