1-Iodo-4-(methylsulfonyl)benzene ≥95%

Supplier: AOBChem USA

Warning

223118-5G 223118-25G 223118-1G 223118-500MG
77634-392EA 609 USD
77634-392 77658-042 77641-280 77649-320
1-Iodo-4-(methylsulfonyl)benzene ≥95%
1-Iodo-4-(methylsulfonyl)benzene

Precaución: For Research Use Only

Formula: C₇H₇IO₂S
MW: 282.099 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00955957
CAS Number: 64984-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR