2,3-Difluoro-4-(trimethylsilyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
22286-5G
22286-25G
22286-1G
22286-500MG
77628-134EA
1484.23
USD
77628-134
77654-230
77641-274
77645-256
2,3-Difluoro-4-(trimethylsilyl)phenylboronic acid ≥97%
2,3-Difluoro-4-(trimethylsilyl)phenylboronic acid
Precaución: For Research Use Only
Formula:
230.092 g/mol MW: C₉H₁₃BF₂O₂Si Storage Temperature: Ambient |
MDL Number:
MFCD22572292 CAS Number: 603990-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...