6-Chloro-2,3-dihydro-7-iodo-1,4-benzodioxin ≥95%

Supplier: AOBChem USA

Warning

222244-5G 222244-500MG 222244-1G
77628-132EA 1484.23 USD
77628-132 77652-498 77641-272
6-Chloro-2,3-dihydro-7-iodo-1,4-benzodioxin ≥95%
6-Chloro-2,3-dihydro-7-iodo-1,4-benzodioxin

Precaución: For Research Use Only

Formula: C₈H₆ClIO₂
Storage Temperature: Ambient
MDL Number: MFCD24115698
CAS Number: 1499313-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR