6-Ethyl-2,3-dihydro-1,4-benzodioxin ≥95%

Supplier: AOBChem USA

Warning

222138-5G 222138-1G 222138-500MG
77654-228EA 2556 USD
77654-228 77641-270 77645-252
6-Ethyl-2,3-dihydro-1,4-benzodioxin ≥95%
6-Ethyl-2,3-dihydro-1,4-benzodioxin

Precaución: For Research Use Only

Formula: 164.2 g/mol
MW: C₁₀H₁₂O₂
Storage Temperature: Ambient
CAS Number: 14939-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR