5-(Methylthio)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

22159-5G 22159-1G 22159-25G 22159-500MG
77632-054EA 803 USD
77632-054 77637-266 77654-220 77649-312
5-(Methylthio)nicotinic acid ≥97%
5-(Methylthio)nicotinic acid

Precaución: For Research Use Only

Formula: C₇H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD01860194
CAS Number: 74470-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR