2-Bromo-4-(trifluoromethoxy)benzoic acid ethyl ester ≥97%
Supplier: AOBChem USA
|
Warning
|
22142-5G
22142-1G
22142-25G
22142-500MG
77632-052EA
933
USD
77632-052
77637-264
77658-036
77645-250
2-Bromo-4-(trifluoromethoxy)benzoic acid ethyl ester ≥97%
2-Bromo-4-(trifluoromethoxy)benzoic acid ethyl ester
Precaución: For Research Use Only
Formula:
313.068 g/mol MW: C₁₀H₈BrF₃O₃ Storage Temperature: Ambient |
MDL Number:
MFCD14698379 CAS Number: 1214337-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...