2-Ethoxy-4-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

22100-5G 22100-500MG 22100-25G 22100-1G
77634-388EA 454 USD
77634-388 77652-496 77654-218 77641-268
2-Ethoxy-4-fluorobenzotrifluoride ≥97%
2-Ethoxy-4-fluorobenzotrifluoride

Precaución: For Research Use Only

Formula: C₉H₈F₄O
Storage Temperature: Ambient
MDL Number: MFCD19105407
CAS Number: 1233541-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR