4-Chloro-7-methoxy-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22097-5G 22097-1G 22097-500MG
77628-128EA 1699.5 USD
77628-128 77637-262 77645-248
4-Chloro-7-methoxy-1H-indazole ≥97%
4-Chloro-7-methoxy-1H-indazole

Precaución: For Research Use Only

Formula: C₈H₇ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09749924
CAS Number: 1000341-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR