1-(Cyclohexylmethyl)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

220930-5G 220930-25G 220930-1G 220930-500MG
77628-126EA 1696.41 USD
77628-126 77658-034 77641-266 77649-310
1-(Cyclohexylmethyl)-4-methylbenzene ≥97%
1-(Cyclohexylmethyl)-4-methylbenzene

Precaución: For Research Use Only

Formula: 188.31 g/mol
MW: C₁₄H₂₀
Storage Temperature: Ambient
MDL Number: MFCD34869111
CAS Number: 92298-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR