4-Methoxydibenzothiophene ≥97%

Supplier: AOBChem USA

Warning

22085-5G 22085-1G 22085-500MG 22085-25G
77632-050EA 1117 USD
77632-050 77637-260 77652-494 77654-216
4-Methoxydibenzothiophene ≥97%
4-Methoxydibenzothiophene

Precaución: For Research Use Only

Formula: C₁₃H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD00092677
CAS Number: 24444-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR