4-[(5-Bromo-2-fluoro-3-pyridinyl)carbonyl]morpholine ≥97%
Supplier: AOBChem USA
|
Warning
|
22063-5G
22063-25G
22063-1G
22063-500MG
77632-048EA
841
USD
77632-048
77654-214
77641-264
77645-246
4-[(5-Bromo-2-fluoro-3-pyridinyl)carbonyl]morpholine ≥97%
4-[(5-Bromo-2-fluoro-3-pyridinyl)carbonyl]morpholine
Precaución: For Research Use Only
Formula:
289.101 g/mol MW: C₁₀H₁₀BrFN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD19687063 CAS Number: 875781-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...