5-Bromo-N,N-diethyl-2-fluoronicotinamide ≥97%

Supplier: AOBChem USA

Warning

22062-5G 22062-1G 22062-25G 22062-500MG
77632-046EA 841 USD
77632-046 77637-258 77658-032 77649-308
5-Bromo-N,N-diethyl-2-fluoronicotinamide ≥97%
5-Bromo-N,N-diethyl-2-fluoronicotinamide

Precaución: For Research Use Only

Formula: 275.117 g/mol
MW: C₁₀H₁₂BrFN₂O
Storage Temperature: Ambient
MDL Number: MFCD19105402
CAS Number: 1010836-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR