1-(2-Chloro-3-fluoropyridin-4-yl)propan-1-one ≥97%

Supplier: AOBChem USA

Warning

220268-5G 220268-1G 220268-25G 220268-500MG
77628-124EA 1484.23 USD
77628-124 77637-256 77658-030 77645-244
1-(2-Chloro-3-fluoropyridin-4-yl)propan-1-one ≥97%
1-(2-Chloro-3-fluoropyridin-4-yl)propan-1-one

Precaución: For Research Use Only

Formula: C₈H₇ClFNO
Storage Temperature: Ambient
MDL Number: MFCD34869107
CAS Number: 2969383-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR