1-(2-Methoxypyridin-4-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

220245-5G 220245-25G 220245-1G 220245-500MG
77632-044EA 1357 USD
77632-044 77654-212 77641-262 77649-306
1-(2-Methoxypyridin-4-yl)propan-1-one ≥95%
1-(2-Methoxypyridin-4-yl)propan-1-one

Precaución: For Research Use Only

Formula: 165.1891 g/mol
MW: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD11656396
CAS Number: 880767-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR