5-Chloro-2-fluoro-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22005-5G 22005-1G 22005-25G 22005-500MG
77632-042EA 656 USD
77632-042 77637-254 77658-028 77649-304
5-Chloro-2-fluoro-3-(trifluoromethyl)benzaldehyde ≥97%
5-Chloro-2-fluoro-3-(trifluoromethyl)benzaldehyde

Precaución: For Research Use Only

Formula: 226.555 g/mol
MW: C₈H₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD26045244
CAS Number: 117752-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR