3-Bromo-5-chloro-2-iodobenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

22002-5G 22002-1G 22002-500MG 22002-25G
77634-386EA 285 USD
77634-386 77637-252 77652-492 77654-210
3-Bromo-5-chloro-2-iodobenzotrifluoride ≥95%
3-Bromo-5-chloro-2-iodobenzotrifluoride

Precaución: For Research Use Only

Formula: C₇H₂BrClF₃I
Storage Temperature: Ambient
MDL Number: MFCD09800701
CAS Number: 1027512-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR