3-Ethoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

21948-5G 21948-1G 21948-500MG
77628-122EA 1484.23 USD
77628-122 77641-260 77649-302
3-Ethoxy-5-(trifluoromethyl)benzoic acid ≥97%
3-Ethoxy-5-(trifluoromethyl)benzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD19105395
CAS Number: 1310416-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR