Cyclopropyl(2,3-difluoro-4-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

219262-5G 219262-1G 219262-500MG
77654-206EA 2073 USD
77654-206 77641-256 77645-240
Cyclopropyl(2,3-difluoro-4-methylphenyl)methanone ≥95%
Cyclopropyl(2,3-difluoro-4-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 196.19 g/mol
MW: C₁₁H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD34149093
CAS Number: 1702212-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR