Cyclopropyl(2,3-difluorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

219209-5G 219209-1G 219209-25G 219209-500MG
77632-040EA 1327 USD
77632-040 77637-248 77654-204 77645-238
Cyclopropyl(2,3-difluorophenyl)methanone ≥97%
Cyclopropyl(2,3-difluorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD16250201
CAS Number: 865664-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR