2-Bromo-4-fluoro-5-methoxybenzonitrile ≥95%

Supplier: AOBChem USA

Warning

218485-5G 218485-25G 218485-1G 218485-500MG
77632-036EA 898 USD
77632-036 77658-024 77641-250 77645-232
2-Bromo-4-fluoro-5-methoxybenzonitrile ≥95%
2-Bromo-4-fluoro-5-methoxybenzonitrile

Precaución: For Research Use Only

Formula: C₈H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD28129639
CAS Number: 1410791-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR