3-Fluoro-5-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

21830-5G 21830-1G 21830-25G 21830-500MG
77634-384EA 475 USD
77634-384 77637-242 77654-200 77645-230
3-Fluoro-5-methoxypyridine ≥97%
3-Fluoro-5-methoxypyridine

Precaución: For Research Use Only

Formula: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD13185606
CAS Number: 1060801-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR